Learn More
Agilent BioTek™ Neurite Outgrowth Module for Gen5 Software
Sfoglia Tutto Agilent BioTek Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
Gen5 Image Prime
Dimensione della confezione:
1 unità
Descrizione
For use with Gen5 Image Prime Software ≥ v3.16 (part number GEN5IPRIME), purchased separately.
- The Agilent BioTek Gen5 neurite outgrowth module accurately quantifies neuronal cell metrics and provides masking options including soma and neurite masks, along with skeletonized neurite masks.
- The Gen5 neurite outgrowth module accurately detects neuronal outgrowth in kinetically monitored, unlabeled live cells.
Module, Software, Neurite, Outgrowth module
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Gen5 Image Prime |
| Tipo | Software |