missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurofascin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26351-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Neurofascin encodes an L1 family immunoglobulin cell adhesion molecule with multiple IGcam and fibronectin domains. The protein functions in neurite outgrowth, neurite fasciculation, and organization of the axon initial segment (AIS) and nodes of Ranvier on axons during early development. Both the AIS and nodes of Ranvier contain high densities of voltage-gated Na+ (Nav) channels which are clustered by interactions with cytoskeletal and scaffolding proteins including this protein, gliomedin, ankyrin 3 (ankyrin-G), and betaIV spectrin. This protein links the AIS extracellular matrix to the intracellular cytoskeleton. Neurofascin undergoes extensive alternative splicing, and the full-length nature of some variants has not been determined.Specifica
| Neurofascin | |
| Polyclonal | |
| Unconjugated | |
| NFASC | |
| KIAA0756, Neurofascin, neurofascin homolog (chicken), NF, NFASC, NRCAML | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116690, 23114, 269116 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O94856, P97685, Q810U3 | |
| NFASC | |
| Neurofascin Fusion Protein Ag24165 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto