missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFIB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16671-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.Specifica
| NFIB | |
| Polyclonal | |
| Unconjugated | |
| NFIB | |
| CTF, HMGIC/NFIB, NF I/B, NF1 B, NFI B, NFI RED, NFIB, NFIB2, NFIB3, Nuclear factor 1 B type, Nuclear factor 1/B, nuclear factor I/B, TGGCA binding protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18028, 29227, 4781 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00712, P97863 | |
| Nfib | |
| NFIB Fusion Protein Ag10165 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto