Learn More
R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit
Codice prodotto. 18700609
Click to view available options
Quantità:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
29 Plex
3 Plex
30 Plex
31 Plex
32 Plex
33 Plex
34 Plex
35 Plex
36 Plex
37 Plex
38 Plex
39 Plex
4 Plex
40 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Dimensione della confezione:
1 set
Pezzo
Codice prodotto. 18700609
Marca: R&D Systems FCSTM2128
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 28 Plex
Specie Primate
Bersaglio legante Multiplex Cytokine Assay Kits
Da utilizzare con (applicazione) ELISA
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.