Learn More
Ansell Nipro GAMMEX™ Polyisoprene Surgical Undergloves
Click to view available options
Size:
5.5
6
6.5
7
7.5
8
8.5
9
Dimensione della confezione:
50 paia
Descrizione
- ASTM D3577, ASTM D6978, ASTM D7160 certified
- Ansell's proprietary soft yet strong formulation delivers heightened comfort and hand protection when double gloving
- Being green in color, these sterile gloves enable fast and easy breach detection when worn with a cream or white outer glove
- Optimal allergy protection; eliminate the risk of developolyisopreneng a latex allergy while skin-friendly formulation helps minimize sensitization to developolyisopreneng a Type IV chemical allergy, as chemical accelerators known to cause skin allergies have been eliminated
- A unique internal coating enables easy donning and doffing under dry or damp hand conditions
- For use in chemotherapy and healthcare applications
Specifica
Specifica
| Certificazione/Conformità | ASTM D3577, ASTM D6978, ASTM D7160, ISO 10282 |
| Certifications/Compliance (Europe) | EN 16523-1, EN 374:2003, EN 420:2003 + A1:2009, EN 455 1-4, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-5:2016, AS/NZS 4179 |
| Accelerator Free | No |
| Applicazioni | Healthcare |
| AQL | 0.65 |
| Colore | Green |
| Monouso | Single-use |
| Finitura esterna | Partially Textured |
| Finger Thickness (English) | 7.87 ±0.79 mil |
| Finger Thickness (Metric) | 0.2 ±0.02 mm |
| Vedi altri risultati |
No evidence of sensitization reported in independent Human Repeat Insult Patch Test (HRIPT).