Learn More
Euro Protection™ Nitrile Gloves, Powder-Free
Click to view available options
Size:
7
8
9
Dimensione della confezione:
pezzi 100
Descrizione
- Liquidproof
- Good dexterity
- Comfort and flexibility
- Mechanical strength
- Suitable for food contact
- Applications: Handling of objects, food industry (preparation), painting, solvent, liquid, assembly of small parts, assembly, cleaning, hospital environments.
Specifica
Specifica
| Certifications/Compliance (Europe) | EN374-1:2016K(6/6)TYPE CEN374-5:2016EN420 |
| Colore | Blue |
| Monouso | Single-use |
| Materiale | Nitrile |
| Powdered | Powder Free |
| Product Type | Nitrile Examination Glove |
| Size | 7 |
| Sterilità | Non-sterile |
| Spessore (sistema metrico) | 0.10 mm |
| Modello | 5916 |
| Vedi altri risultati |