Learn More
Euro Protection™ Nitrile Gloves, Powder-Free
Codice prodotto. p-7146805
Click to view available options
Size:
7
8
9
Dimensione della confezione:
pezzi 100
Codice prodotto. 15392286
Marca: Euro Protection™ 5918
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Euro Protection™ Nitrile Gloves, Powder-Free provide comfort, flexibility, and dexterity for a variety of tasks including food preparation.
  • Liquidproof
  • Good dexterity
  • Comfort and flexibility
  • Mechanical strength
  • Suitable for food contact
  • Applications: Handling of objects, food industry (preparation), painting, solvent, liquid, assembly of small parts, assembly, cleaning, hospital environments.
Specifica
Certifications/Compliance (Europe) EN374-1:2016K(6/6)TYPE CEN374-5:2016EN420
Colore Blue
Monouso Single-use
Materiale Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size 8
Sterilità Non-sterile
Spessore (sistema metrico) 0.10 mm
Modello 5918
Quantità 100/Pk.