Learn More
Aalborg™ Nitrogen Direct Flow Tube 150mm Reference Scale
Click to view available options
Velocità di flusso dell'aria:
100 mL/min.
2 L/min.
200 mL/min.
27 L/min.
300 mL/min.
500 mL/min.
Dimensione della confezione:
Pezzo
Descrizione
- Direct reading tubes are straightforward.
- The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
- Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
- Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifica
Specifica
| Tipo | Nitrogen Direct Flow Tube |
| Larghezza (sistema metrico) | 150 mm |
| Velocità di flusso dell'aria | 27 L/min. |