Learn More
Aalborg™ Nitrogen Direct Flow Tube 150mm Reference Scale
Codice prodotto. p-8490009
Click to view available options
Velocità di flusso dell'aria:
100 mL/min.
2 L/min.
200 mL/min.
27 L/min.
300 mL/min.
500 mL/min.
Dimensione della confezione:
Pezzo
Codice prodotto. 11718288
Marca: Aalborg™ FT03222CAVN
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Aalborg™ Nitrogen Direct Flow Tube 150mm Reference Scale completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifica
Tipo Nitrogen Direct Flow Tube
Larghezza (sistema metrico) 150 mm
Velocità di flusso dell'aria 200 mL/min.