missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKp30/NCR3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Marca: R&D Systems AF1849-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
NKp30/NCR3 Polyclonal specifically detects NKp30/NCR3 in Human samples. It is validated for Western Blot.Specifica
| NKp30/NCR3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activating natural killer receptor p30, CD337, CD337 antigen, CD337activating NK-A1 receptor, LY117,1C7NK-p30, lymphocyte antigen 117, natural cytotoxicity triggering receptor 3, Natural killer cell p30-related protein, NCR3, NKP30, NKp30MALS | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 259197 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q05D23 | |
| NCR3 | |
| Mouse myeloma cell line NS0-derived recombinant human NKp30/NCR3 Leu19-Thr138 Accession # Q05D23 | |
| 25 μg | |
| Primary | |
| Detects human NKp30/NCR3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) NKp80, rhNKp44 and rhNKp46 is observed. | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Fisher Scientific Encompass-programmet erbjuder artiklar som inte ingår i vår distributionsportfölj. Dessa produkter har vanligtvis inga bilder eller detaljerade beskrivningar. Vi är dock angelägna om att förbättra din shoppingupplevelse. Använd formuläret nedan för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering