missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOD-IN-1, MedChemExpress
Click to view available options
Quantità:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Description
NOD-IN-1 is a potent mixed inhibitor of nucleotide-binding oligomerization domain (NOD)-like receptors, NOD1 and NOD2, with IC50 of 5.74 μM and 6.45 μM, respectively.
Specifications
Specifications
| Materiale o nome chimico | NOD-IN-1 |
| CAS | 132819-92-2 |
| Colore | White |
| Note sul grado di purezza | Research |
| Formula molecolare | C18H17NO4S |
| Quantità | 50 mg |
| Informazioni di solubilità | DMSO : 100 mg/mL (291.21 mM; Need ultrasonic) |
| SMILES | O=S(N1C(C(OCC)=O)=CC2=CC=CC=C21)(C3=CC=C(C)C=C3)=O |
| Molecular Weight (g/mol) | 343.4 |
| Peso formulazione | 343.4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction