Learn More
Standard di torbidità non ratio
BIOPHARMA
Codice prodotto. p-8009959 Sfoglia Tutto Reagecon Prodotti
Click to view available options
Volume (sistema metrico):
100 ml
500 mL
Torbidità:
0.0 NTU
1 NTU
10 NTU
100 NTU
20 NTU
200 NTU
40 NTU
4000 NTU
5 NTU
50 NTU
500 NTU
800 NTU
Dimensione della confezione:
100mL
500mL
Codice prodotto. 11738505
Marca: Reagecon CRS10100
Use turbidity standards for precise measurements with nephelometeric instruments. Non Ratio Turbidity Standards, Reagecon™ are composed of suspended polymer microspheres to eliminate the problems with Formazin standards. They are NIST traceable, US EPA approved and certified accurate to ±1%.
These turbidity standards remove the handling, stability and accuracy problems associated with traditional Formazin turbidity standards. They are certified traceable to NIST Reference Material 1690, and are available in a range of concentrations from 0–4000 NTU.
  • Non-toxic and non-carcinogenic, with no special handling or disposal requirements
  • Well defined spherical shape of particles, 0.01–0.2μm in diameter
  • Particles stay in suspension—they do not deteriorate or settle out
  • A long stable shelf life of 2 years at all concentrations
  • Convenient, ready-to-use solution—mixing is discouraged as it entrains air
Specifica
Tipo Standard di torbidità non ratio
Da utilizzare con (apparecchiatura) Strumenti non ratio
Da utilizzare con (applicazione) Nephelometric turbidity measurement
Accuracy ±1%
Torbidità 10 NTU
Certificazione/Conformità NIST, approvazione US EPA
Volume (sistema metrico) 100 ml