Learn More
Emma Safety Footwear Nordic Oiled Nubuck Leather D-Fit Shoes
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
Dimensione della confezione:
1 paio
Pezzo
Descrizione
- Oil Nubuck uppers
- Hydro-Tec™ sanitized silver lining
- Non-woven antistatic insole
- GripForce™ Easy Twist outsole
Specifica
Specifica
| Colore | Black, Brown |
| Materiale rivestimento interno | Hydro-Tec sanitized silver |
| Materiale | Leather |
| Size | 43 |
| Caratteristiche suola | Non-woven antistatic insole |
| Materiale suola | GripForce™ Easy Twist outsole |
| Materiale dello zoccolo | Steel |
| Product Type | Oiled Nubuck Leather Shoes |