Learn More
Normal Mouse IgM, Alexa Fluor 700, Negative Control, R&D Systems™
Mouse Polyclonal Antibody
Marca: R&D Systems IC015N
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgM Polyclonal specifically detects IgM in Mouse samples. It is validated for Control.
Specifica
| IgM | |
| Alexa Fluor 700 | |
| AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
| Mouse | |
| Protein A or G purified | |
| 3507 | |
| Mouse | |
| IgM |
| Polyclonal | |
| Control | |
| IGHM | |
| NA | |
| 200 Tests | |
| Serum was obtained from naive (non-immunized) mice and purified for use as a mouse IgM control for flow cytometry applications. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |