missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notch-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-27347
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Notch-2 Polyclonal specifically detects Notch-2 in Human samples. It is validated for Western Blot.
Specifica
| Notch-2 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| NOTCH2 | |
| A portion of amino acids 1550-1600 of humanNOTCH2 was used as the immunogen. | |
| 0.1 mg | |
| Apoptosis, Cancer, Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Signal Transduction, Stem Cell Signaling Pathway | |
| 4853 | |
| Human | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 2-4ug/ml | |
| AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, 94% homologous in pig and horse, and 88% homologous in mouse, rat, dog and bovine. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto