Learn More
Emma Safety Footwear Novara Safety Shoes
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Dimensione della confezione:
1 paio
Descrizione
- S3-Safety shoe with sole plate for wet work
- Full grain leather / Nubuck construction
- Chemical resistant / Liquid repellant
- Nitrile Rubber Heat Resistant Outsole
- ESD certified
- Chrome-free lining
- Sanitized
Specifica
Specifica
| Certificazione/Conformità | EN ISO 20345:2011, EN 12568:2010 |
| Tipo di chiusura | Laces |
| Colore | Brown |
| Progettazione | Novara |
| Materiale rivestimento interno | Hydro-Tec™ Sanitized Silver & Wellness leather |
| Materiale | Leather, Nubuck |
| Size | 43 |
| Materiale dello zoccolo | Steel |