missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human/Monkey Extracellular Protein Buffer Magnetic Reagent Kit

Codice prodotto. 13406436
Click to view available options
Quantità:
Each
Dimensione della confezione:
Pezzo
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 13406436

Marca: Invitrogen™ LHB0001M

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Includes

Contains diluents, SAV concentrate, buffers, wash solution, magnetic plate, filter plate and complete protocol.

Use with Human and Monkey extracellular magnetic Singleplex kits for in vitro quantitative determination of analytes in serum, plasma, and tissue culture supernatant

Singleplex magnetic bead kits maximize flexibility in experimental design, permitting the measurement of one or multiple analytes in panels designed by the researcher.

Luminex xMAP™ Technology—Effective and Proven Analytical Tool
Human and Monkey extracellular magnetic singleplex kits are based on xMAP; technology. The use of a suspension bead-based technology enables the multiplexing capabilities of the Luminex assays. Magnetic microspheres are internally dyed with red and infrared fluorophores of differing intensities. Each bead is given a unique number, or bead region, allowing differentiation of one bead from another. Beads covalently bound to different antibodies can be mixed in the same assay, utilizing a 96-well microplate format. At the completion of the sandwich immunoassay, beads can be read using the Luminex 200 or FLEXMAP 3D detection systems, instruments based on flow cytometry, two lasers, real-time digital signal processing, and xPONENT™ software to distinguish bead color (analyte) and assay signal strength (PE) fluorescence intensity.

MAGPIX™ System—Brings the Power of Multiplex to Every Lab
The MAGPIX system may also be used for sample analysis. This instrument utilizes xMAP technology and is built on light-emitting diodes (LEDs) for excitation, a CCD camera for bead and analyte detection and high-speed digital signal processors. The MAGPIX system is more compact and robust, providing a very cost effective magnetic bead-based multiplexing tool. Simple, out of the box set-up allows researchers to rapidly perform singleplex or multiplex assays of up to 50 targets in single biological samples. The MAGPIX system is easy to use—ideal for both new and experienced users alike.

Magnetic Beads Let You Do More with Your Time
Magnetic bead-based assays use Luminex MagPlex magnetic microsphere. All other assay components are the same as the equivalent Luminex polystyrene bead-based assays providing identical quality and consistency performance. MagPlex technology takes advantage of magnetic properties to simplify assay wash steps and maximize uniformity of results. Magnetic bead-based assays also enable automation, decrease hands-on time, and increase throughput and precision. Easy to use protocols allow you to get your results in as little as 3.5 hours. Assays are compatible with both vacuum and magnetic wash stations and work well with the MAGPIX system as well as other Luminex xMAP platforms.

More Validation for Confidence in Your Results
Human and Monkey extracellular magnetic singleplex kits have been validated using the same rigorous criteria that distinguish Luminex assays from other commercially available assays. Every singleplex kit and premixed multiplex kit comes with a product insert that outlines the specifications for each marker in the assay. Reagents suitable for use with Luminex 200™ FLEXMAP 3D™, and MAGPIX™ systems.

Cell Analysis, Immunoassays, Ready-To-Use Immunoassay

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifica

Quantità Each
Specie Human, Monkey
Da utilizzare con (apparecchiatura) Luminex 200 System, MAGPIX System, FlexMAP 3D System
Linea di prodotti Novex
Content And Storage Contains diluents, SAV concentrate, buffers, wash solution, magnetic plate, filter plate and complete protocol. Store at 2-8° C.
Shipping Condition Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato