missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Acinus Antibody Blocking Peptide
Sfoglia Tutto Bio Techne Prodotti
Click to view available options
Quantità:
0.05 mg
Dimensione della confezione:
0.05mg
Descrizione
A blocking peptide containing 15 amino acids near the center of human Acinus. Source: Synthetic (Accession #: AAD56724) The Acinus Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifica
Specifica
| Gene ID (Entrez) | 22985 |
| Specie | Human |
| Content And Storage | Store at -20 C. Avoid freeze-thaw cycles. |
| Formulazione | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. |
| Da utilizzare con (applicazione) | This Peptide is Useful as a Blocking Peptide for NBP1-76855. For Further Blocking Peptide Related Protocol, Click Here . |
| Alias gene | Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107 |
| Simbolo del gene | ACIN1 |
| Product Type | Acinus |
| Quantità | 0.05 mg |
| Status giuridico | RUO - research use only |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction