missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Noxa Antibody (114C307.1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB600-1159F
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Noxa Monoclonal specifically detects Noxa in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifica
| Noxa | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| PMAIP1 | |
| This Noxa antibody was developed by immunizing mice with a fusion protein containing human Noxa. | |
| Protein G purified | |
| RUO | |
| Primary | |
| An ∼80 kD background band may also be seen in Jurkat cell lysate. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 114C307.1 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa | |
| Mouse | |
| 14.96 kDa | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Markers, Membrane Trafficking and Chaperones, Necroptosis, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors | |
| 5366 | |
| Human, Mouse, Rat | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto