Learn More
NRAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10724-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
NRAS (GTPase NRas) is a membrane protein that shuttles between the Golgi apparatus and the plasma membrane. This shuttling is regulated through palmitoylation and depalmitoylation by the ZDHHC9-GOLGA7 complex. This protein, which has intrinsic GTPase activity, is activated to a GTP-bound form by a GTPase activating protein and inactivated to a GDP-bound form by a guanine nucleotide-exchange factor. Defects in the gene encoding this protein are a cause of juvenile myelomonocytic leukemia (JMML) and Noo syndrome 6.Specifica
| NRAS | |
| Polyclonal | |
| Unconjugated | |
| NRAS | |
| ALPS4, GTPase NRas, HRAS1, N ras, NRAS, NRAS1, Transforming protein N Ras | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18176, 24605, 4893 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P01111, P08556, Q04970 | |
| Nras | |
| NRAS Fusion Protein Ag1081 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |