Learn More
NSUN2 Mouse anti-Human, CoraLite 488, Clone: 2H5A2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66580-100UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The NSUN2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NSUN2. This antibody recognizes human antigen. The NSUN2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifica
| NSUN2 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 54888 | |
| IgG2a |
| Immunofluorescence | |
| 2H5A2 | |
| Mouse | |
| Primary | |
| Human |