missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 8 Trace, 8-well kit for DNA from forensic samples

Codice prodotto. p-9281008
Click to view available options
N. di reazioni:
480 Preps
96 Preps
Dimensione della confezione:
96 reazioni
Pezzo
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 12708552

Marca: Macherey-Nagel™ 740722.1

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Isolation of DNA from forensic samples in flexible 8‐well strip format.

Lysis is achieved by incubation of samples in a solution containing chaotropic ions in the presence of proteinase K. If subsequent separation of the lysate from the sample material is difficult, the lysate can be separated from the forensic material under vacuum or centrifugation by use of the NucleoSpin™ Trace filter plate (not included in the kits). Appropriate conditions for binding of DNA to the silica membrane in the NucleoSpin™ Trace binding strip/binding plate are created by addition of isopropanol to the lysate. The binding process is reversible and specific to nucleic acids. Contaminants are removed by two washing steps with ethanolic buffer. Pure genomic DNA is finally eluted under low ionic strength conditions in a slightly alkaline elution buffer.Typically yields of 1 to 2 μg genomic DNA can be purified from buccal swabs. The final concentration of eluted DNA is 10 to 20 ng/μL (depending on elution buffer volume). Typically, A260/280 is 1.8 to 1.9. The obtained DNA can be used directly as template for PCR.

Kit components

NucleoSpin™ Trace binding strips, MN wash plates, MN square well blocks, rack with tube strips, cap strips, self-adhering foil, buffers, proteinase K. NucleoSpin™ 8 Trace - NucleoSpin™ Trace binding plates, MN wash plates, MN square well blocks, racks with tube strips, cap strips, buffers, proteinase K.

  • Suitable for manual and automated processing, processing possible under vacuum or by centrifugation
  • 8 well strip format
  • Typical yield: 1 to 2 μg DNA from buccal swabs
  • Elution volume: 50 to 100 μL
  • Silica membrane technology
  • No use of organic solvents
  • Highly pure nucleic acids suitable for all common downstream applications
  • Preparation time: 30 min./8 strip
  • Binding capacity: 20 μg

Specifica

Content And Storage 18°C to 25°C
Formato 8-well Strip
Da utilizzare con (applicazione) Isolation of DNA
N. di reazioni 96 Preps
compliance-icons compliance-icons
Identificativo prodotto
  • NucleoSpin 8 Trace (12x8)
Avvertenza
  • Pericolo
Categoria di pericolo
  • Lesione oculare grave/irritazione oculare Categoria 2
  • Sensibilizzante delle vie respiratorie Categoria 1
  • Corrosione/irritazione della pelle Categoria 2
  • Tossicità specifica per organi bersaglio (esposizione singola) Categoria 3
Indicazioni di pericolo
  • H315-Provoca irritazione cutanea.
  • H319-Provoca grave irritazione oculare.
  • H334-Può provocare sintomi allergici o asmatici o difficoltà respiratorie se inalato.
  • H335-Può irritare le vie respiratorie.
Consigli di prudenza
  • P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
  • P342+P311-In caso di sintomi respiratori: contattare un CENTRO ANTIVELENI/un medico/ .
Informazioni supplementari
  • MIXTURE LIST-Contiene: proteinase K
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato