Learn More
Thermo Scientific™ Nunc™ Cell Factory™ Systems with Active Gassing
Click to view available options
Area di coltura:
25,280 cm2
2528 cm²
6320 cm²
Livelli vassoi:
4
10
40
Dimensione della confezione:
pezzi 10
pezzi 2
pezzi 6
Descrizione
Create an ideal environment for culturing oxygen-consuming and pH sensitive cells using Thermo Scientific™ Nunc™ Cell Factory™ Systems with Active Gassing. Flow system helps ensure a controlled, contamination-free atmosphere in each culture tray by equally distributing the user-specified gas mixture. Available in 4-, 10- and 40-tray models.
- Achieve greater yields with controlled cell culture environments equally distributed within each layer
- Optimize production capacity without changing cell culture platforms
- Develop industrial scale solutions with the utilization of an active gassing Cell Factory™ Incubator
Specifica
Specifica
| Descrizione | Cell Factory with Active Gassing |
| N. per custodia | 10 |
| N. per confezione | 10 |
| Product Type | System with Active Gassing |
| Sterilità | Sterile |
| Area di coltura | 2528 cm² |
| Livelli vassoi | 4 |
| Quantità | Pack/Case: 1/10 |