Learn More
Thermo Scientific™ Nunc™ Lab-Tek™ Petri Dishes
Click to view available options
Area di coltura:
79 sq. cm
Dimensione della confezione:
pezzi 25
Descrizione
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc™ Lab-Tek™ Petri Dishes.
- Available with a molded grid
- Performs well in automatic dispensers
- Contact dish useful for sampling in hospital environments as well as in food science and the pharmaceutical industry
- Compatible with automated systems
Specifica
Specifica
| Descrizione | Vented, without stack ring |
| Forma del fondo | Piatto |
| Area di coltura | 79 sq. cm |
| Forma | Square |
| Tipo | Piastra di Petri |
| Conteggio pozzetti | 1 |
| Sfiato | Yes |