Learn More
Masterflex™ Nylon Female Luer Fitting
Click to view available options
Da utilizzare con:
Female Luer x 1/8 in. Wide Bore Hose Barb
Female Luer x 1/8-27 NPT Elbow
Female Luer x 3/32 in. Hose Barb
Female Luer x 3/32 in. Wide Bore Hose Barb
Dimensione della confezione:
pezzi 25
Descrizione
- Standard hose barb for use with flexible tubing
- Smooth bore hose barb to reduce flow turbulence and risk of line contamination
- Wide-bore hose barb for enhanced fluid path flow
Specifica
Specifica
| Materiale | Nylon |
| Da utilizzare con | Female Luer x 1/8-27 NPT Elbow |
| Quantità confezione | 25 Pack |
| Product Type | Female Luer Fitting |