Learn More
Cole-Parmer™ Essentials Disposable Forceps, Reinforced Nylon, Serrated Pointed Tips
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Lunghezza (sistema metrico):
114 mm
Dimensione della confezione:
pezzi 100
Descrizione
- Corrugated handles provide you with a secure grip.
- Reduce cross contamination with these disposable forceps.
- Disposable.
- Forceps are available in Polystyrene and reinforced Nylon.
- Choose from sterile, autoclavable, as well as blunt or pointed tips.
- Sterile forceps are individually wrapped with easy to open peel-packs.
- Non-sterile forceps are autoclavable.
- Serrated pointed tips are ideal for handling smaller objects.
- Serrated blunt tips are more for general use.
Specifica
Specifica
| Stile punta | Pointed |
| N. per confezione | 100 Pk. |
| Lunghezza (sistema imperiale) | 4.5 in. |
| Materiale | Nylon |
| Lunghezza (sistema metrico) | 114 mm |
| Product Type | Forceps |
| Quantità (N. per custodia) | 100 Pk. |
| Tipo | Forceps |