Learn More
DWK Life Sciences O-Ring, red, FEP coated, not suitable for desiccators
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Diametro (sistema metrico) parte esterna:
75 mm
110 mm
133 mm
157 mm
215 mm
Dimensione della confezione:
Pezzo
Descrizione
Flat flange vessels are large-volume reaction vessels consisting of a lower part and a lid. These two parts are held together by a clamping ring. The red O-ring made of silicone is seamlessly coated with FEP and serves as an optimal seal for flat flange vessels with an annular groove. It prevents jamming of the ground surfaces between the base and the lid and is convincing due to its good elasticity in combination with outstanding chemical resistance.
- Accessories for flat flange vessels, comprising an elastic, silicone core with seamless FEP coating that encloses the ring
- The combination of these high-quality materials achieves good elasticity in conjunction with outstanding chemical resistance
Specifica
Specifica
| Da utilizzare con (applicazione) | Tight closure of flange vessels |
| Quantità | 1 Ea. |
| Materiale | Silicone, FEP |
| Diametro (sistema metrico) parte esterna | 75 mm |