Learn More
Octadecyltriethoxysilane, 98%, n-isomer 85% min
Product Code. 11362486
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362486 50 g 50g
11352486 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11362486 Supplier Thermo Scientific Alfa Aesar Supplier No. L06366.18
 Restricted Item
In stock
Add to basket
CAS: 7399-00-0 | C24H52O3Si | 416.76 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7399-00-0
Molecular Formula C24H52O3Si
Molecular Weight (g/mol) 416.76
MDL Number MFCD00039875
InChI Key FZMJEGJVKFTGMU-UHFFFAOYSA-N
Synonym octadecyltriethoxysilane, triethoxy octadecyl silane, n-octadecyltriethoxysilane, octadecyl triethoxysilane, stearyl triethoxysilane, unii-1vn9p25l8h, triethoxystearylsilane, acmc-209ot8
PubChem CID 81885
IUPAC Name triethoxy(octadecyl)silane
SMILES CCCCCCCCCCCCCCCCCC[Si](OCC)(OCC)OCC
Specifications
Boiling Point 165°C to 169°C (0.2mmHg)
Quantity 50 g
Flash Point 195°C (383°F)
Refractive Index 1.438
Sensitivity Moisture sensitive
Density 0.87
Beilstein 7025163
Formula Weight 416.77
Percent Purity 98%
Assay n-isomer ≥85%
Chemical Name or Material Octadecyltriethoxysilane