Learn More
Ohaus™ OIML F1 Weight Set
Sfoglia Tutto Ohaus Prodotti
Click to view available options
Massa:
1 kg to 1 g
1 kg to 1 mg
100 g to 1 g
100 g to 1 mg
2 x 2 kg to 1 g
2 x 2 kg to 1 mg
2 x 200 g to 1 g
2 x 200 g to 1 mg
5 kg to 1 g
5 kg to 1 mg
50 g to 1 g
50 g to 1 mg
500 g to 1 g
500 g to 1 mg
500 mg to 1 mg
Dimensione della confezione:
Pezzo
Descrizione
- Weights Sets in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free to protect your investment
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifica
Specifica
| Certificazione/Conformità | No Certificate |
| Materiale | Stainless Steel, Aluminum |
| Peso (sistema metrico) | 0.431 kg |
| Massa | 500 mg to 1 mg |
| Include | Protective Case |
| Quantità confezione | 12/Pk. |