Learn More
Oleic Acid N-Hydroxysuccinimide, TRC
Codice prodotto. 17915015
Cambia vista
Click to view available options
Quantità:
100 mg
1 g
Dimensione della confezione:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
17915015 100 mg 100mg
17921715 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 17915015 Fornitore Toronto Research Chemicals N. del fornitore TRCO524300100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 81480-40-2
Formula molecolare C22 H37 N O4
Molecular Weight (g/mol) 379.53
Sinonimo (9Z)-9-Octadecenoic Acid 2,5-Dioxo-1-pyrrolidinyl Ester, (Z)-1-[(1-Oxo-9-octadecenyl)oxy]-2,5-pyrrolidinedione, 1-[[(9Z)-1-Oxo-9-octadecenyl]oxy]-2,5-pyrrolidinedione, N-Hydroxysuccinimide Ester with Oleic Acid, N-Hydroxysuccinimide Oleate, N-Succinimidyl Oleate, Oleic Acid N-Hydroxysuccinimide Ester, Succinimidyl Oleate
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (Z)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)ON1C(=O)CCC1=O
Specifica
Materiale o nome chimico Oleic Acid N-Hydroxysuccinimide
Quantità 100 mg
InChI Formula InChI=1S/C22H37NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-22(26)27-23-20(24)18-19-21(23)25/h9-10H,2-8,11-19H2,1H3/b10-9-
Peso formulazione 379.2723