Learn More
Cytiva Omega Membrane Discs
Click to view available options
MWCO:
1 kDa
10 kDa
100 kDa
30 kDa
5 kDa
Diametro (sistema metrico) membrana:
43 mm
62 mm
76 mm
Dimensione della confezione:
pezzi 12
Descrizione
- Polyethersulfone membrane provides higher flow rates and lower protein binding than competitive regenerated cellulose
- Lower processing times and the highest possible recoveries
- Discs can be washed and reused
- Replacement membranes for other stirred cell systems concentration, fractionation, purification, buffer exchange, or desalting of protein, cell broths, and other biomolecules
- For research use only
Specifica
Specifica
| Materiale (membrana) | PES |
| MWCO | 30 kDa |
| Sterilità | Non-sterile |
| N. per confezione | 12/Pk. |
| Diametro (sistema metrico) membrana | 76 mm |