Learn More
Diba Omnifit™ EZ Chromatography Columns with One Fixed and One Adjustable Endpieces
Codice prodotto. 15815404
Encompass_Preferred
Click to view available options
Lunghezza (sistema metrico):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Diametro (sistema metrico) interno:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Capacità della colonna:
0.1 to 0.7 mL
0.1 to 2.4 mL
0.2 to 5.5 mL
0.4 to 12.4 mL
1.2 to 34.4 mL
1.4 to 4.1 mL
108.0 to 147.3 mL
11.0 to 17.3 mL
134.7 to 211.7 mL
142.4 to 181.6 mL
17.3 to 23.6 mL
19.6 to 58.9 mL
191.4 to 230.7 mL
211.7 to 288.6 mL
22.8 to 29.1 mL
24.7 to 38.9 mL
279.0 to 356.0 mL
280.4 to 440.7 mL
3.1 to 9.4 mL
30.6 to 36.9 mL
38.5 to 115.5 mL
38.9 to 53.0 mL
4.8 to 7.5 mL
440.7 to 600.9 mL
51.2 to 65.4 mL
580.9 to 741.1 mL
68.7 to 108.0 mL
68.9 to 83.1 mL
7.1 to 21.2 mL
7.5 to 10.3 mL
781.2 to 941.4 mL
9.9 to 12.7 mL
missing translation for 'unitSize'
Pezzo
Product Code. 15815404
missing translation for 'mfr': Diba 006EZ3533AF
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Includes
Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting.
Omnifit™ EZ Chromatography Columns with One Fixed and One Adjustable Endpieces are ergonomically designed endpieces provide simple and quick height adjustment
  • PEEK™ column threads offer outstanding durability
  • Precision borosilicate glass columns provide inertness, visibility, and performance
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Specifications
Intervallo di temperatura (sistema metrico) 4 to 40°C
Diametro (sistema metrico) interno 35 mm
Tipo Liquid Chromatography Column
Pressione max 150 psi
Dimensione del poro 10 and 30 μm
Capacità della colonna 211.7 to 288.6 mL
Lunghezza (sistema metrico) 330 mm
Materiale Borosilicate Glass, PTFE, Polyethylene, FKM/FPM
Temperatura max 40°C
Include Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF PP fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits), and a Click-N-Seal™ 10-32 PEEK conical port fitting.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.