Learn More
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Adjustable Endpieces
Codice prodotto. 15885414
Encompass_Preferred
Click to view available options
Lunghezza (sistema metrico):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Diametro (sistema metrico) interno:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Capacità della colonna:
0.1 to 0.7 mL
0.1 to 2.4 mL
0.1 to 4.1 mL
0.2 to 5.5 mL
0.2 to 9.4 mL
0.4 to 12.4 mL
0.4 to 21.2 mL
1.2 to 34.4 mL
1.2 to 58.9 mL
10.6 to 38.9 mL
103.1 to 181.6 mL
11 to 23.6 mL
120.2 to 440.7 mL
134.7 to 288.6 mL
152.2 to 230.7 mL
16.5 to 29.1 mL
2.1 to 7.5 mL
2.3 to 115.5 mL
202.0 to 356.0 mL
24.3 to 36.9 mL
24.7 to 53.0 mL
27.7 to 211.7 mL
280.4 to 600.9 mL
29.5 to 108.0 mL
37.1 to 65.4 mL
4.7 to 17.3 mL
4.8 to 10.3 mL
420.6 to 741.1 mL
54.8 to 83.1 mL
620.9 to 941.4 mL
68.7 to 147.3 mL
7.2 to 12.7 mL
missing translation for 'unitSize'
Pezzo
Product Code. 15885414
missing translation for 'mfr': Diba 006EZS0605AA
Only null left
Add to basket
Add to basket
Omnifit™ EZ SolventPlus™ Chromatography Columns with One Fixed and One Adjustable Endpieces
  • PEEK™ column threads offer outstanding durability
  • Ergonomically designed adjustable endpiece offers quick and easy height adjustment
  • Superior frit placement adds bed support and even distribution of sample flow
  • Reusable columns let you pack your own media specific to your application
Specifications
Intervallo di temperatura (sistema metrico) 4 to 40°C
Diametro (sistema metrico) interno 6.6 mm
Tipo Liquid Chromatography Column
Pressione max 900 psi
Dimensione del poro 10 and 30 μm
Capacità della colonna 0.1 to 0.7 mL
Lunghezza (sistema metrico) 50 mm
Materiale Borosilicate Glass, PTFE, Chemraz™
Temperatura max 40°C
Include Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.