Learn More
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Fixed Endpieces
Click to view available options
Lunghezza (sistema metrico):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
440 mm
500 mm
Diametro (sistema metrico) interno:
6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Capacità della colonna:
0.7 mL
2.4 mL
4.1 mL
5.5 mL
7.5 mL
9.4 mL
10.3 mL
12.4 mL
12.7 mL
17.3 mL
21.2 mL
23.6 mL
29.1 mL
34.4 mL
36.9 mL
38.9 mL
53 mL
58.9 mL
65.4 mL
83.1 mL
108 mL
115.5 mL
147.3 mL
181.6 mL
211.7 mL
230.7 mL
288.6 mL
356 mL
440.7 mL
600.9 mL
741.1 mL
941.4 mL
Unit Size:
Pezzo
Description
- PEEK™ column threads offer outstanding durability
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Specifications
Specifications
| Intervallo di temperatura (sistema metrico) | 4 to 40°C |
| Diametro (sistema metrico) interno | 50 mm |
| Tipo | Liquid Chromatography Column |
| Pressione max | 75 psi |
| Dimensione del poro | 10 and 30 μm |
| Capacità della colonna | 440.7 mL |
| Lunghezza (sistema metrico) | 250 mm |
| Materiale | Borosilicate Glass, PTFE, Chemraz™ |
| Temperatura max | 40°C |
| Include | Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.