Learn More
OPA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 27733-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
OPA1 is a dynamin-related GTPase that is critical for the maintece of mitochondrial morphology and mtDNA. The most commonly associated phenotype with OPA1 mutations is heterozygous optic atrophy, a heterozygous domit trait that causes reduced visual clarity and sometimes blindness. The disease usually begins in childhood and increases in severity throughout the life of affected individuals. Usually, this phenotype is attributed to the degeneration of optic nerve fibers. Interestingly, the same type of nerve degeneration seems to be partially causative of certain schizophrenia characteristics. OPA1 dysfunction also seems to be implicated in this case; mitochondrial networks associated with critical nerves seem to link schizophrenia and OPA1. The dysfunction is associated with issues with apoptosis and normal cellular metabolic regulation, all regulated through OPA1.Specifica
| OPA1 | |
| Polyclonal | |
| Unconjugated | |
| OPA1 | |
| KIAA0567, largeG, MGM1, NTG, OPA1, Optic atrophy protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171116, 4976, 74143 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60313, P58281, Q2TA68 | |
| Opa1 | |
| OPA1 Fusion Protein Ag26887 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |