Learn More
OR2T3/OR2T34 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca:  Invitrogen PA538224
Codice prodotto. 15535151
  • 486.16€ / 100µg
Questo articolo non è restituibile. Consulta la politica di reso
Description
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OR2T3/OR2T34
Polyclonal
Unconjugated
OR2T3
OR2T3, OR2T34
A synthetic peptide derived from the C-terminal region of human OR2T3/2T34
100 μg
Primary
Human
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Q8NGX1, Q8NH03
Rabbit
Antigen affinity chromatography
RUO
127068, 343173
-20°C
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.