missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR52E1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA538178
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifica
| OR52E1 | |
| Polyclonal | |
| Unconjugated | |
| OR52E1 | |
| OR52E1P | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79296 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8NGJ3 | |
| OR52E1 | |
| A synthetic peptide derived from the internal region of human OR52E1 | |
| 100 μg | |
| Primary | |
| Human, Non-human Primate | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto