Learn More
Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits
Descrizione
Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.
- For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
- No. of tests: 50 to 100
Specifica
Specifica
| Tipo | Chlorine |
| Formati reagenti test | Tablet |
| Da utilizzare con (apparecchiatura) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast |
| Descrizione | AQUAfast chlorine (free & total) reagent tablets, 50 tests each |
| Include | Set of 2: 50 DPD No 1 Tablets, 50 DPD No 3 Tablets |
| Metodo | DPD |
| Da utilizzare con (applicazione) | Colorimetric water and wastewater testing equipment |
| Accessori raccomandati | AQUAfast 24mm round vials (Cat. No. AC2V24) |
| Range | 0.01 to 6 mg/L Cl2 |
| Metodi approvati EPA | Drinking Water: Standard Methods 4500-Cl G; Wastewater: Standard Methods 4500-Cl G-2000 |
| Vedi altri risultati |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.