Learn More
Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor
Codice prodotto. 10140731
Click to view available options
Materiale o nome chimico:
Low Ionic Strength pH Test Kit
pH Buffer A
pH Buffer B
pH Buffer C
pHISA Low Ionic Strength Adjustor
Dimensione della confezione:
Pezzo
pezzi 4
pezzi 5
Codice prodotto. 10140731
Marca: Thermo Scientific™ 700001
Sample well, rain, distilled/deionized, boiler feed and process waters with this adjustor.
Sample well water, rain water, distilled/deionized water, boiler feed water, process water and more with the Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor. It offers fast, accurate pH results in high purity and low ionic strength waters.
Features:
  • Increases measurement accuracy
  • Reduces electrode drift
  • Eliminates contamination of sample
Pure Water pH Test Kit:
  • Kit contains special low ionic strength buffers for calibration and pHISA™ adjustor additive, to increase sample ionic strength without changing its pH
  • For best results, a high quality pH electrode such as the Thermo Scientific™ Orion™ ROSS Ultra™ glass-body pH electrode, Cat. No. 8102BNUWP, is recommended
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Low Ionic Strength pH Test Kit
Da utilizzare con (applicazione) Provide fast, accurate pH results in high purity and low ionic strength waters. Recommended for samples such as well water, rain water, distilled/deionized water, boiler feed water and process water.
Da utilizzare con (apparecchiatura) pH Meter and Electrode
pH 6.97
Quantità 4 x 475 mL pH 6.97 Buffer, 2 x 475 mL pH 4.10 Buffer, 2 x 60 mL pHISA Adjuster
Imballaggio Kit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.