Learn More
OTC Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551366
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: ILADYLTLQE HYSSLKGLTL SWIGDGNNIL HSIMMSAAKF GMHLQAATPK GYEPDASVTK LAEQYAKENG TKLLLTNDPL EAAHGGNVLI TDTWISMGQE EEKKKRLQAF QGYQVTMKTA KVAASDWT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also.Specifica
OTC | |
Polyclonal | |
Unconjugated | |
OTC | |
AI265390; OCTD; ornithine carbamoyltransferase; ornithine carbamoyltransferase, mitochondrial; Ornithine transcarbamylase; Otc; OTCase; Sf; sparse fur; spf | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5009 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P00480 | |
OTC | |
Recombinant protein corresponding to Human OTC | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only