Learn More
Chemsplash Overshoes (Pairs) 5 and 6, Anti-Slip Sole, Elasticated Ankle
GREENER_CHOICE
Codice prodotto. p-200105900
Click to view available options
Size:
Medium
Large
X-Large
Dimensione della confezione:
200 paia
Codice prodotto. 30058349
Marca: Chemsplash 2564.400.005
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Anti-Slip Overshoe is made from Chemsplash fabric which is soft and microporous. It is strong and resistant to chemical splash and dust.
Chemsplash Anti-Slip Overshoe
  • Chemsplash 55GSM Type PB 6B Material
  • Coated White PVC Slip-Resistant Sole
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • 16 in. in Length
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
  • PFAS Free
Description: The Chemsplash Anti-Slip Overshoe is made from Chemsplash fabric which is soft and microporous. It is strong and resistant to chemical splash and dust. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. An anti-slip textured sole has been added to enhance grip with the floor, and an elasticated ankle ensures a secure fit. Chemsplash accessories can be used to protect the wearer from their environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Specifica
Size Medium
Materiale Microporous Fabric
Latex Free Yes
Colore White
Monouso Single-use
Sterilità Non-sterile
Tipo di chiusura Elastic Ankle
Tipo di suola Anti-slip
Resistenza Liquid
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 100-1000 (ISO 5-6)