Learn More
Chemsplash Overshoes (Pairs) PB 6B Elasticated Ankle
GREENER_CHOICE
Codice prodotto. p-200105899
Click to view available options
Size:
Large
X-Large
Dimensione della confezione:
200 paia
Codice prodotto. 30058391
Marca: Chemsplash 2560.400.006
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Elasticated Overshoe is made from Chemsplash soft and microporous fabric. It is strong and resistant to chemical splash and dust.
Chemsplash Elasticated Overshoe
  • Chemsplash 55GSM Type PB 6B Material
  • 16 in. in Length
  • Silicone and Latex Free
  • Non-Linting
  • Anti-Static Material
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
  • PFAS Free
Description: The Chemsplash Elasticated Overshoe is made from Chemsplash soft and microporous fabric. It is strong and resistant to chemical splash and dust. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories can be used to protect the wearer from their environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Specifica
Size Large
Materiale Microporous Fabric
Latex Free Yes
Colore White
Monouso Single-use
Sterilità Non-sterile
Tipo di chiusura Elastic Ankle
Tipo di suola Seamless
Resistenza Liquid
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 100-1000 (ISO 5-6)