Learn More
Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems
Click to view available options
Volume (sistema metrico) tampone:
300 mL
550 mL
1250 mL
Include:
Apparatus Only
Complete System
Complete System plus Caster
Dimensione della confezione:
Pezzo
Descrizione
Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.
Simple, Six-Step Use:
- Secure the glass cassettes
- Pour the agarose
- Plug in the specially designed trough
- Cast polyacrylamide solution in the cassettes
- Load samples
- Run the gel
Features:
- Manufactured using a time-tested acrylic fabrication technology, providing a durable, long-lasting system
- Central cooling core provides even cooling across the gel surface for outstanding resolution and clear banding
- Cooling is achieved by circulating fluid through the core with external circulator or cool tap water
- Very simple clamping mechanism makes loading glass cassettes and most manufacturers' precast gels trouble free
Recommended for:
- SDS electrophoresis
- The second dimension of two-dimensional (2-D) electrophoresis
- Agarose electrophoresis
- Native electrophoresis
Specifica
Specifica
| Product Line | Owl |
| Tipo | Dual-Gel Vertical Electrophoresis System Apparatus |
| Include | Apparatus Only |
| Modello | P9DS |
| Volume (sistema metrico) tampone | 550 mL |
| Lunghezza (sistema imperiale) | 9.8 in. |
| Lunghezza (sistema metrico) gel | 14 to 16 cm |
| Larghezza (sistema imperiale) | 5.1 in. |
| Larghezza (sistema metrico) gel | 16 cm |
| Altezza (sistema imperiale) | 7.7 in. |