Learn More
Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs
Click to view available options
Tipo:
Double-sided Comb
Prep Comb
Rapid Load Comb
Standard Comb
Conteggio denti:
10
12
14
17 (2X)
2
5
8
9 (1X)
Spessore (sistema metrico) denti:
1 mm
1.0/1.5 mm
1.5 mm
Dimensione della confezione:
Pezzo
Descrizione
Eliminate comb assembly with Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis System Combs. These heavy-duty one-piece combs are for use with the EasyCast™ B1 Mini Gel Electrophoresis System.
Heavy-duty, one-piece combs eliminate comb assembly.
Ordering Information:
Cat Nos. B1-Rl-9C, -9D, -17C and -17D are 8 and 12-channel pipette format (1X) and (2X) combs.
Customized combs are available (Order SKU # XCM). Please contact your Customer Service or Sales Representative for further details.
Specifica
Specifica
| Descrizione | Double-sided 10 tooth comb; 1.0/1.5mm |
| Da utilizzare con (apparecchiatura) | Owl™ EasyCast B1 Mini Gel Electrophoresis System |
| Gel Size | Mini |
| Product Line | Owl, EasyCast |
| Tipo | Double-sided Comb |
| Conteggio denti | 10 |
| Spessore (sistema metrico) denti | 1.0/1.5 mm |