missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OX40 Ligand (CD252) Mouse anti-Human, BV605, Clone: ik-1, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 743234
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
OX40 Ligand (CD252) Monoclonal antibody specifically detects OX40 Ligand (CD252) in Human samples. It is validated for Flow Cytometry
Specifica
| OX40 Ligand (CD252) | |
| Monoclonal | |
| Brilliant Violet 605 | |
| CD252; OX40 Ligand; CD134 Ligand; CD134L; TNFSF4; GP34; TXGP1 | |
| Human OX40 Ligand Transfected Cell Line | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| ik-1 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto