Learn More
Thermo Scientific™ Oxoid™ Blaser-Wang Campylobacter Selective Supplement
Click to view available options
Quantità:
10/Pk.
Dimensione della confezione:
pezzi 10
Descrizione
Oxoid Campylobacter Selective Supplement, Blaser-Wang is used to prepare of Blaser-Wang Selective Medium for the isolation of Campylobacter while inhibiting growth of Candida albicans.
- Add to Columbia Blood Agar Base, Part No. CM0271B
- Each vial supplements 500mL of medium
Specifica
Specifica
| Descrizione | Campylobacter Selective Supplement, Blaser-Wang |
| Forma | Freeze-dried |
| Product Type | Selective Supplement |
| Quantità | 10/Pk. |
| Resa | Each vial for 500mL medium |
| Specie | Campylobacter |