Learn More
Thermo Scientific™ Oxoid™ Gentamicin Selective Supplement
Click to view available options
Quantità:
10 x 256 mg
Dimensione della confezione:
pezzi 10
Descrizione
Oxoid Gentamicin Selective Supplement (256mg/vial) is used for the isolation of Vancomycin-Resistant Enterococci (VRE) and High Level Aminoglycoside-Resistant Enterococci (HLARE) from clinical specimens.
- Add to VRE Agar Base, Part No. CM0985B
- Each vial supplements 500mL of medium
Specifica
Specifica
| Descrizione | Gentamicin Selective Supplement |
| Forma | Freeze-dried |
| Product Type | Selective Supplement |
| Quantità | 10 x 256 mg |