Learn More
Thermo Scientific™ Oxoid™ Malt Extract
Codice prodotto. p-200112597
Click to view available options
Quantità:
Confezioni singole
Dimensione della confezione:
500g
Codice prodotto. 10188572
Marca: Thermo Scientific™ LP0039B
Oxoid Malt Extract is recommended for use in media for yeast and mold growth.
An extract of sprouted grain with conserved nitrogen and carbohydrate constituents
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Disidratato; estratto di malto
Forma In polvere
Product Type Estratto di malto
Quantità Confezioni singole
compliance-icons
Product Identifier
  • Malt Extract
Signal Word
  • Pericolo
Hazard Category
  • Sensibilizzante delle vie respiratorie Categoria 1
Hazard Statement
  • H334-Può provocare sintomi allergici o asmatici o difficoltà respiratorie se inalato.
Precautionary Statement
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P261-Evitare di respirare la polvere/i fumi/i gas/la nebbia/i vapori/gli aerosol.
  • P304-IN CASO DI INALAZIONE:.
  • P342+P311-In caso di sintomi respiratori: contattare un CENTRO ANTIVELENI/un medico/ .
Supplemental information
  • MIXTURE LIST-Contiene: Barley, malt
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.