Learn More
Thermo Scientific™ Oxoid™ Meropenem Selective Supplement
Click to view available options
Quantità:
10/Pk.
Dimensione della confezione:
pezzi 10
Descrizione
Oxoid Meropenem Selective Supplement (1mg/vial) is used for the isolation of Vancomycin-Resistant Enterococci (VRE) and HighLevel Aminoglycoside-Resistant Enterococci (HLARE) from clinical specimens.
- Add to VRE Agar Base, Part No. CM0984B
- Each vial supplements 500mL of medium
Specifica
Specifica
| Descrizione | Meropenem Selective Supplement |
| Forma | Freeze-dried |
| Product Type | Selective Supplement |
| Quantità | 10/Pk. |
| Resa | Each vial for 500mL medium |