Learn More
Thermo Scientific™ Oxoid™ Mupirocin Antimicrobial Susceptibility discs
Codice prodotto. p-6148777
Click to view available options
Concentrazione:
200 μg
5 μg
Dimensione della confezione:
pezzi 250
Codice prodotto. 11933822
Marca: Thermo Scientific™ CT0522B
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Determine the susceptibility of microorganisms using manual AST methods and a full range of high quality, easy-to-use antimicrobial susceptibility testing discs.
Manually determine the antibiotic susceptibility of microorganisms using Antimicrobial Susceptibility Testing (AST) methods in conjunction with reliable, easy-to-use Thermo Scientific™ Oxoid™ Mupirocin Antimicrobial Susceptibility discs.
Ease of use is provided by shatterproof spring-loaded cartridges, easy-to-read labeling and end-of-cartridge indicators.
disc diffusion not only categorizes resistant, intermediate and sensitive organisms through quantitative results, but also provides a visual indication of the following:
  • Inoculum level
  • Presence of contamination
  • Resistant mutants
  • Beta-lactamase activity
  • Antagonism and synergy between antibiotics
Each cartridge is individually sealed with a desiccant to maintain a moisture level of <2%, ensuring long term stability.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifica
Agenti antimicrobici Mupirocin
Certificazione/Conformità Complies with BSAC, SFM guidelines
Concentrazione 5 μg
Descrizione Oxoid Mupirocin Antimicrobial Susceptibility Discs
Codice antibiotico MUP
Product Type Antimicrobial Susceptibility Discs
Test Type Antimicrobial Susceptibility
N. per confezione 5 Cartridges/Pk.
Tipo di imballaggio 50 Discs/Cartridge
Quantità 50 Discs/Cartridge